Ethylsorbat ≥97%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
Sorbinsäureethylester, Ethyl-(2E,4E)-2,4-hexadienoat, (2E,4E)-2,4-Hexadiensäureethylester, Ethyl-trans,trans-2,4-hexadienoat, 2,4-Hexadiensäureethylester, Ethyl-(E,E)-2,4-hexadienoat, (E,E)-2,4-Hexadiensäureethylester, trans,trans-2,4-Hexadiensäureethylester
A11431.14
A11431.22
A11431.36
A11431.14EA
24.3
EUR
A11431.14
A11431.22
A11431.36
Ethylsorbat ≥97%
Ethylsorbat
Purity: 98%
CAS No.: 2396-84-1
Molecular Formula: CH3CH=CHCH=CHCO2CH2CH3
Formula Weight: 140.18
Flash Point: 69°(156°F)
Refractive Index: 1.4950
Boiling Point: 195-196°
Beilstein Reference: 1362941
MDL No.: MFCD00009296
Formel:
C₈H₁₂O₂ Molecular Weight: 140,18 g/mol Siedepunkt: 195…196 °C Dichte: 0,948 Flammpunkt: 69 °C (156 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009296 CAS-Nummer: 2396-84-1 EINECS: 219-258-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...