1-Brom-2-methoxyethan
Lieferant: FLUOROCHEM
Synonyme:
(2-Bromethyl)methylether
005433-25G
005433-100G
FLUO005433-25GEA
29.9
EUR
FLUO005433-25G
FLUO005433-100G
1-Brom-2-methoxyethan
1-Brom-2-methoxyethan
Formel:
BrH₂CCH₂OCH₃ Molecular Weight: 138.99 g/mol Siedepunkt: 110 Dichte: 1.46 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000236 CAS-Nummer: 6482-24-2 EINECS: 229-339-4 UN: 3271 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...