8-Brom-1-octanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Octamethylenbromhydrin, 1-Brom-8-hydroxyoctan
294144-1G
294144-5G
SIAL294144-1GEA
43.2
EUR
SIAL294144-1G
SIAL294144-5G
8-Brom-1-octanol, Sigma-Aldrich®
8-Brom-1-octanol
Formel:
BrH₂C(CH₂)₆CH₂OH Molecular Weight: 209,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010388 CAS-Nummer: 50816-19-8 EINECS: 256-785-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...