Trichloracetonitril 98

Lieferant: Thermo Fisher Scientific
Gefahr

A10565.0B A10565.18 A10565.30
A10565.0BEA 487 EUR
A10565.0B A10565.18 A10565.30
Trichloracetonitril 98
Trichloracetonitril

Purity: 98% CAS No.: 545-06-2 Molecular Formula: C2Cl3N Formula Weight: 144.39 Physical Form: Clear liquid Density: 1.44 Refractive Index: 1.4410 Appearance: Colourless Melting Point: -44° Boiling Point: 85-86° Beilstein Reference: 605572 Merck Reference: 14,9628 UN No.: 3276 MDL No.: MFCD00001842

Formel: C₂Cl₃N
Molecular Weight: 144.39 g/mol
Siedepunkt: 85…86 °C
Schmelzpunkt: –44 °C
Dichte: 1.44
Lagertemperatur: Raumtemperatur
MDL: MFCD00001842
CAS-Nummer: 545-06-2
EINECS: 208-885-7
UN: 3276
Merck Index: 12,09757

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR