2,2-Dichloracetamid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L00315.09 L00315.18
L00315.09EA 37 EUR
L00315.09 L00315.18
2,2-Dichloracetamid ≥98%
2,2-Dichloracetamid

Purity: 98+% CAS No.: 683-72-7 Molecular Formula: C2H3Cl2NO Formula Weight: 127.96 Melting Point: 96-101° Boiling Point: 233-234° MDL No.: MFCD00008015

Formel: C₂H₃Cl₂NO
Siedepunkt: 233…234 °C
Schmelzpunkt: 96…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008015
CAS-Nummer: 683-72-7
EINECS: 211-674-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR