1-Chlor-4-iodbutan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

396222-25ML 396222-5ML
SIAL396222-25MLEA 265 EUR
SIAL396222-25ML SIAL396222-5ML
1-Chlor-4-iodbutan, Sigma-Aldrich®
1-Chlor-4-iodbutan
Formel: C₄H₈ClI
Molecular Weight: 218,46 g/mol
MDL: MFCD00039415
CAS-Nummer: 10297-05-9
EINECS: 233-669-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR