2-Chlorethylisothiocyanat
Lieferant: FLUOROCHEM
002532.10G
002532-1G
002532-5G
002532-25G
FLUO002532.10GEA
0
EUR
FLUO002532.10G
FLUO002532-1G
FLUO002532-5G
FLUO002532-25G
2-Chlorethylisothiocyanat
2-Chlorethylisothiocyanat
Formel:
C₃H₄ClNS Siedepunkt: 80 °C (13 mmHg) Dichte: 1,265 g/cm³ (20 °C) |
MDL:
MFCD00041125 CAS-Nummer: 6099-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...