3-Methoxypropiononitril ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Methoxypropanenitrile, 3-Methoxypropionitrile

L07055.AE L07055.AP L07055.PI
L07055.AEEA 28 EUR
L07055.AE L07055.AP L07055.PI
3-Methoxypropiononitril ≥99%
3-Methoxypropiononitril

Purity: 99% CAS No.: 110-67-8 Molecular Formula: CH3O(CH2)2CN Formula Weight: 85.11 Flash Point: 66°(150°F) Refractive Index: 1.4030 Odour: Weak pH: 6.8 Melting Point: -63° Boiling Point: 164-165° Beilstein Reference: 1739284 MDL No.: MFCD00001958

Formel: C₄H₇NO
Molecular Weight: 85,11 g/mol
Siedepunkt: 164…165 °C
Schmelzpunkt: –63 °C
Dichte: 0,938
Flammpunkt: 66 °C (150 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001958
CAS-Nummer: 110-67-8
EINECS: 203-790-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR