2-Oxoglutarsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Ketoglutarsäure, α-Ketoglutarsäure
A10256.14
A10256.22
A10256.36
A10256.14EA
25.4
EUR
A10256.14
A10256.22
A10256.36
2-Oxoglutarsäure ≥98%
2-Oxoglutarsäure
Purity: 98%
CAS No.: 328-50-7
Molecular Formula: C5H6O5
Formula Weight: 146.10
Storage Temperature: 2°C to 8°C
Melting Point: 113-117°
Beilstein Reference: 1705689
Merck Reference: 14,5303
MDL No.: MFCD00004165
Formel:
HOOCCH₂CH₂COCOOH Molecular Weight: 146,1 g/mol Siedepunkt: 323 °C (1013 hPa) Schmelzpunkt: 113…117 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004165 CAS-Nummer: 328-50-7 EINECS: 206-330-3 Merck Index: 14,5303 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...