4-(Methylthio)butanon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methylthio-2-butanone

M-Clarity™ Qualitätsstufe = MQ300

W337501-100G-K W337501-25G-K
SIALW337501-100G-KEA 833 EUR
SIALW337501-100G-K SIALW337501-25G-K
4-(Methylthio)butanon, Sigma-Aldrich®
4-(Methylthio)butanon
Formel: C₅H₁₀OS
Molecular Weight: 118,2 g/mol
MDL: MFCD00040029
CAS-Nummer: 34047-39-7
EINECS: 251-810-8
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR