Acetamid ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethanamid, Essigsäureamid

A12589.0B A12589.0I A12589.30
A12589.0BEA 88.2 EUR
A12589.0B A12589.0I A12589.30
Acetamid ≥99%
Acetamid

Purity: 99% CAS No.: 60-35-5 Molecular Formula: CH3CONH2 Formula Weight: 59.07 Melting Point: 76-81° Boiling Point: 220-222° Merck Reference: 14,43 MDL No.: MFCD00008023

Formel: C₂H₅NO
Molecular Weight: 59.07 g/mol
Siedepunkt: 220…222 °C
Schmelzpunkt: 76…81 °C
Dichte: 1.159
Lagertemperatur: Raumtemperatur
MDL: MFCD00008023
CAS-Nummer: 60-35-5
EINECS: 200-473-5
Merck Index: 13,00044

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR