Harnstoff 99,0-100,5% ACS
Lieferant: Thermo Fisher Scientific
Synonyme:
Urea, Aminoameisensäureamid, Carbamid, Carbonyldiamid
36428.22
36428.36
36428.A3
36428.22EA
35.6
EUR
36428.22
36428.36
36428.A3
Harnstoff 99,0-100,5% ACS
Harnstoff
Purity: 99.0-100.5%
CAS No.: 57-13-6
Molecular Formula: NH2CONH2
Formula Weight: 60.06
Physical Form: Crystalline
Density: 1.335
pH: 9,2-9,5
Melting Point: 132-135°
Solubility: Soluble in water, alcohol, methanol, glycerol, benzene, and conc. HCl. Practically insoluble in chloroform, ether
Beilstein Reference: 635724
Merck Reference: 14,9867
MDL No.: MFCD00008022
Used as a protein denaturant
Formel:
NH₂CONH₂ Siedepunkt: 197 °C (1013 hPa) Schmelzpunkt: 132…135 °C Dichte: 1,335 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008022 CAS-Nummer: 57-13-6 EINECS: 200-315-5 REACH: 01-2119463277-33 Merck Index: 14,9867 |