N-(2-Acetamido)-2-aminoethansulfonsäure (ACES), Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(Carbamoylmethyl)taurin

M-Clarity™ Qualitätsstufe = MQ200

A9758-100G A9758-1KG A9758-250G A9758-25G
SIALA9758-100GEA 370 EUR
SIALA9758-100G SIALA9758-1KG SIALA9758-250G SIALA9758-25G
N-(2-Acetamido)-2-aminoethansulfonsäure (ACES), Sigma-Aldrich®
N-(2-Acetamido)-2-aminoethansulfonsäure (ACES)
Formel: H₂NCOCH₂NHCH₂CH₂SO₃H
Molecular Weight: 182,2 g/mol
Schmelzpunkt: 270…295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008030
CAS-Nummer: 7365-82-4
EINECS: 230-908-4
Merck Index: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=a5fb2045198b642d124372f31d901db2e2d25597&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR