Acetohydrazid 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Essigsäurehydrazid, Acethydrazid

A10969.14 A10969.22 A10969.36
A10969.14EA 45.1 EUR
A10969.14 A10969.22 A10969.36
Acetohydrazid 96%
Acetohydrazid

Purity: 96% CAS No.: 1068-57-1 Molecular Formula: CH3CONHNH2 Formula Weight: 74.08 Flash Point: 110°(230°F) Melting Point: ca 60-68° Boiling Point: 128-129°/18mm Beilstein Reference: 506165 MDL No.: MFCD00007610

Formel: C₂H₆N₂O
Molecular Weight: 74,08 g/mol
Siedepunkt: 128…129 °C (18 mmHg)
Schmelzpunkt: ∼ 60…68 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007610
CAS-Nummer: 1068-57-1
EINECS: 213-948-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR