Adipohydrazid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Adipyl hydrazide
A15119.14
A15119.22
A15119.36
A15119.14EA
32
EUR
A15119.14
A15119.22
A15119.36
Adipohydrazid ≥97%
Adipohydrazid
Purity: 97%
CAS No.: 1071-93-8
Molecular Formula: H2NNHCO(CH2)4CONHNH2
Formula Weight: 174.20
Odour: Odourless
Melting Point: 180-182°
Beilstein Reference: 973863
MDL No.: MFCD00007614
Formel:
C₆H₁₄N₄O₂ Molecular Weight: 174,2 g/mol Siedepunkt: 446 °C (1013 hPa) Schmelzpunkt: 180…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007614 CAS-Nummer: 1071-93-8 EINECS: 213-999-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...