3'-Nitroacetanilid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(3-Nitrophenyl)acetamid

B22062.06 B22062.14 B22062.22
B22062.06EA 41.6 EUR
B22062.06 B22062.14 B22062.22
3'-Nitroacetanilid ≥98%
3'-Nitroacetanilid

Purity: 98+% CAS No.: 122-28-1 Molecular Formula: C8H8N2O3 Formula Weight: 180.16 Melting Point: 151-153° Beilstein Reference: 2211961 Merck Reference: 14,6581 MDL No.: MFCD00017015

Formel: C₈H₈N₂O₃
Molecular Weight: 180,16 g/mol
Schmelzpunkt: 151…153 °C
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00017015
CAS-Nummer: 122-28-1
EINECS: 204-532-6
Merck Index: 14,6581

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR