4'-Nitroacetanilid 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-(4-Nitrophenyl)acetamid, Ethansäure(4-nitrophenyl)amid
415750250
415751000
ACRO415750250EA
71.1
EUR
ACRO415750250
ACRO415751000
4'-Nitroacetanilid 99%
4'-Nitroacetanilid
Formel:
C₈H₈N₂O₃ Siedepunkt: 409 °C (1013 hPa) Schmelzpunkt: 213…216 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007303 CAS-Nummer: 104-04-1 EINECS: 203-169-0 Merck Index: 12,06677 |
Spezifikation Testergebnisse
Appearance | Yellow to yellow-green to green-brown Solid |
Infrared spectrum | Conforms |
Melting point | 213 to 216 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...