2'-Hydroxyacetanilid ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethansäure(2-hydroxyphenyl)amid, N-Acetyl-2-aminophenol, N-(2-Hydroxyphenyl)acetamid, 2-Acetamidophenol

B21264.14 B21264.22
B21264.14EA 29.1 EUR
B21264.14 B21264.22
2'-Hydroxyacetanilid ≥97%
2'-Hydroxyacetanilid

Purity: 97% CAS No.: 614-80-2 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 205-210° Beilstein Reference: 607592

Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
Schmelzpunkt: 205…210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002181
CAS-Nummer: 614-80-2
EINECS: 210-396-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR