2'-Hydroxyacetanilid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethansäure(2-hydroxyphenyl)amid, N-Acetyl-2-aminophenol, N-(2-Hydroxyphenyl)acetamid, 2-Acetamidophenol

M-Clarity™ Qualitätsstufe = MQ200

A7000-100G-A A7000-25G-A
SIALA7000-100G-AEA 94.3 EUR
SIALA7000-100G-A SIALA7000-25G-A
2'-Hydroxyacetanilid, Sigma-Aldrich®
2'-Hydroxyacetanilid
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00002181
CAS-Nummer: 614-80-2
EINECS: 210-396-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR