Butylstearat, tech.
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Stearinsäurebutylester, Butyloctadecanoat
36338.36
36338.A3
36338.36EA
57.5
EUR
36338.36
36338.A3
Butylstearat, tech.
Butylstearat
CAS No.: 123-95-5
Molecular Formula: CH3(CH2)16COO(CH2)3CH3
Formula Weight: 340.59
Flash Point: 160°(320°F)
Physical Form: Liquid
Density: 0.861
Refractive Index: 1.4430
Appearance: Colourless to pale yellow
Melting Point: 17-22°
Boiling Point: 343°
Beilstein Reference: 1792866
Merck Reference: 14,1589
MDL No.: MFCD00026669
Formel:
C₂₂H₄₄O₂ Molecular Weight: 340.59 g/mol Siedepunkt: 343 °C Schmelzpunkt: 17…22 °C Dichte: 0.861 Flammpunkt: 160 °C (320 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026669 CAS-Nummer: 123-95-5 EINECS: 204-666-5 Merck Index: 14,1589 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...