Methylpropionat ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Propanoic acid methyl ester

A14944.0F A14944.AE A14944.AP
A14944.0FEA 343 EUR
A14944.0F A14944.AE A14944.AP
Methylpropionat ≥99%
Methylpropionat

Purity: 99% CAS No.: 554-12-1 Molecular Formula: CH3CH2CO2CH3 Formula Weight: 88.11 Flash Point: -2°(28°F) Physical Form: Liquid Density: 0.915 Refractive Index: 1.3770 Melting Point: -88° Boiling Point: 78-80° Beilstein Reference: 1737628 Merck Reference: 14,6112 UN No.: 1248 MDL No.: MFCD00009306

Formel: C₄H₈O₂
Molecular Weight: 88.11 g/mol
Siedepunkt: 78…80 °C
Schmelzpunkt: –88 °C
Dichte: 0.915
Flammpunkt: –2 °C (28 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009306
CAS-Nummer: 554-12-1
EINECS: 209-060-4
UN: 1248
Merck Index: 12,06191

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR