Dimethyladipat ≥99%

Lieferant: Thermo Scientific

Synonyme: Adipic acid dimethyl ester, 1,6-Hexanedioic acid dimethyl ester

B21174.0B B21174.18 B21174.30
B21174.0BEA 80 EUR
B21174.0B B21174.18 B21174.30
Dimethyladipat ≥99%
Dimethyladipat

Purity: 99% CAS No.: 627-93-0 Molecular Formula: CH3O2C(CH2)4CO2CH3 Formula Weight: 174.20 Flash Point: 107°(224°F) Refractive Index: 1.4280 Melting Point: 8° Boiling Point: 109-110°/14mm Beilstein Reference: 1707443 Merck Reference: 14,162 MDL No.: MFCD00008469

Formel: C₈H₁₄O₄
Molecular Weight: 174,2 g/mol
Siedepunkt: 109…110 °C (14 mmHg)
Schmelzpunkt: 8 °C
Dichte: 1,063
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008469
CAS-Nummer: 627-93-0
EINECS: 211-020-6
Merck Index: 14,162

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR