Kaliumformiat, (max. 2% H₂O) 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ameisensäure Kaliumsalz

A14551.30 A14551.0B A14551.0I
A14551.30EA 33.2 EUR
A14551.30 A14551.0B A14551.0I
Kaliumformiat, (max. 2% H₂O) 99%
Kaliumformiat

Purity: 99% CAS No.: 590-29-4 Molecular Formula: HCO2K Formula Weight: 84.12 Melting Point: 165-168° Solubility: Soluble in water Beilstein Reference: 3623272 Merck Reference: 14,7633 MDL No.: MFCD00013100

Formel: CHKO₂
Molecular Weight: 84,12 g/mol
Schmelzpunkt: 165…168 °C
Dichte: 1,91
Lagertemperatur: Raumtemperatur
MDL: MFCD00013100
CAS-Nummer: 590-29-4
EINECS: 209-677-9
Merck Index: 14,7633

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR