Bismuttriacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Bismut(III)acetat, Essigsäure Bismut(III)salz
M-Clarity™ Qualitätsstufe = MQ100
401587-10G
401587-50G
SIAL401587-10GEA
92.7
EUR
SIAL401587-10G
SIAL401587-50G
Bismuttriacetat, Sigma-Aldrich®
Bismuttriacetat
Formel:
(H₃CCOO)₃Bi Molecular Weight: 386,11 g/mol |
MDL:
MFCD00015629 CAS-Nummer: 22306-37-2 EINECS: 244-904-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...