5-Aminovaleriansäure 97%
Lieferant: Thermo Scientific
Synonyme:
5-Aminopentansäure
153910050
153910250
ACRO153910050EA
47.6
EUR
ACRO153910050
ACRO153910250
5-Aminovaleriansäure 97%
5-Aminovaleriansäure
Formel:
C₅H₁₁NO₂ Molecular Weight: 117,15 g/mol Siedepunkt: 248 °C (1013 hPa) Schmelzpunkt: 158…161 °C Dichte: 1,33 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008232 CAS-Nummer: 660-88-8 EINECS: 211-544-5 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Solubility | (5 % in water) Clear colorless to light yellow solution |
Titration with HClO4 | 96.0 to 104.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...