D-Methionin 99+%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-(+)-2-Amino-4-(methylmercapto)buttersäure, (R)-(+)-Methionin, (R)-(+)-2-Amino-4-(methylthio)buttersäure, D(+)-Methionin, H-D-Met-OH

227210050 227210250 227211000
ACRO227210050EA 42.1 EUR
ACRO227210050 ACRO227210250 ACRO227211000
D-Methionin 99+%
D-Methionin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149.21 g/mol
Siedepunkt: 278
Schmelzpunkt: 273…275 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00002622
CAS-Nummer: 348-67-4
EINECS: 206-483-6
Merck Index: 13,06004

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Sulfated ash ≤0.1 %
UV transmittance ≥98 % (c=3, H2O, 430 nm) 1 cm cell
Specific optical rotation -23° to -25° (20°C, 589 nm) (c=8, 6 N HCl) (on dried substance)
Titration with HClO4 99.0 to 101.0 % (on dried substance)
Chloride (Cl) ≤300 ppm
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids (TLC) not detectable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR