1,4-Difluorbutan

Lieferant: FLUOROCHEM

010297-1G 010297-5G
FLUO010297-1GEA 320 EUR
FLUO010297-1G FLUO010297-5G
1,4-Difluorbutan
1,4-Difluorobutane
Formel: C₄H₈F₂
MDL: MFCD01740905
CAS-Nummer: 372-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR