3'-Fluoracetanilid
Lieferant: FLUOROCHEM
Synonyme:
N-(3-Fluorphenyl)acetamid
006174-25G
006174-100G
FLUO006174-25GEA
36.9
EUR
FLUO006174-25G
FLUO006174-100G
3'-Fluoracetanilid
3'-Fluoracetanilid
Formel:
C₈H₈FNO Molecular Weight: 153.16 g/mol |
MDL:
MFCD00017917 CAS-Nummer: 351-28-0 EINECS: 206-509-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...