4'-(Trifluormethyl)acetanilid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-(4-(Trifluormethyl)phenyl)acetamid, Ethansäure(4-(trifluormethyl)phenyl)amid, 4-Acetamidobenzotrifluorid

A12789.06 A12789.14
A12789.06EA 62.8 EUR
A12789.06 A12789.14
4'-(Trifluormethyl)acetanilid ≥98%
4'-(Trifluormethyl)acetanilid

Purity: 98+% CAS No.: 349-97-3 Molecular Formula: C9H8F3NO Formula Weight: 203.16 Odour: Odourless Melting Point: 150-154° Beilstein Reference: 2106965 MDL No.: MFCD00013562

Formel: C₉H₈F₃NO
Molecular Weight: 203.16 g/mol
Schmelzpunkt: 150…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013562
CAS-Nummer: 349-97-3
EINECS: 206-495-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR