4'-Fluoracetanilid

Lieferant: CHEMPUR

Synonyme: Ethansäure(4-fluorphenyl)amid, N-(4-Fluorphenyl)acetamid

FL3844.25 FL3844.100
CHMPFL3844.25EA 0 EUR
CHMPFL3844.25 CHMPFL3844.100
4'-Fluoracetanilid
4'-Fluoracetanilid
Formel: C₈H₈FNO
Molecular Weight: 153,16 g/mol
Siedepunkt: 310 °C (1013 hPa)
Schmelzpunkt: 154 °C
MDL: MFCD00016335
CAS-Nummer: 351-83-7
EINECS: 206-515-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR