Ethyldifluoracetat

Lieferant: FLUOROCHEM

Synonyme: Difluoroacetic ethyl ester

003219-25G 003219-100G 003219-5G
FLUO003219-25GEA 60.9 EUR
FLUO003219-25G FLUO003219-100G FLUO003219-5G
Ethyldifluoracetat
Ethyldifluoracetat
Formel: C₄H₆F₂O₂
Molecular Weight: 124,09 g/mol
Siedepunkt: 98 °C (1013 hPa)
Dichte: 1,1893 g/cm³ (9.8 °C)
MDL: MFCD00013578
CAS-Nummer: 454-31-9
EINECS: 207-223-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR