Methyltrifluoracetat ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Trifluoroacetic methyl ester
A17338.0B
A17338.18
A17338.30
A17338.0BEA
636
EUR
A17338.0B
A17338.18
A17338.30
Methyltrifluoracetat ≥98%
Methyltrifluoracetat
Purity: 98+%
CAS No.: 431-47-0
Molecular Formula: CF3CO2CH3
Formula Weight: 128.05
Flash Point: -18°(-0.4°F)
Refractive Index: 1.2900
Odour: Pungent
Melting Point: -78°
Boiling Point: 43-44°
Beilstein Reference: 1756070
UN No.: 2924
MDL No.: MFCD00000417
Formel:
C₃H₃F₃O₂ Molecular Weight: 128,05 g/mol Siedepunkt: 43…44 °C Schmelzpunkt: –78 °C Dichte: 1,283 Flammpunkt: –18 °C (–0,4 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000417 CAS-Nummer: 431-47-0 EINECS: 207-074-5 UN: 2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...