Methyltrifluoracetat
Lieferant: FLUOROCHEM
Synonyme:
Trifluoroacetic methyl ester
001026-100G
001026-500G
001026-1KG
FLUO001026-100GEA
36.9
EUR
FLUO001026-100G
FLUO001026-500G
FLUO001026-1KG
Methyltrifluoracetat
Methyltrifluoracetat
Formel:
C₃H₃F₃O₂ Molecular Weight: 128.05 g/mol Siedepunkt: 43 Schmelzpunkt: –78 °C Dichte: 1.2639 g/cm³ (20 °C) |
MDL:
MFCD00000417 CAS-Nummer: 431-47-0 EINECS: 207-074-5 UN: 2924 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...