Phenyltrifluoracetat
Lieferant: FLUOROCHEM
Synonyme:
Trifluoroacetic phenyl ester
001215-5G
001215-25G
001215-100G
FLUO001215-5GEA
42.9
EUR
FLUO001215-5G
FLUO001215-25G
FLUO001215-100G
Phenyltrifluoracetat
Phenyltrifluoracetat
Formel:
C₈H₅F₃O₂ Molecular Weight: 190.12 g/mol Siedepunkt: 145…147 °C (1013 hPa) Schmelzpunkt: –8.5 °C Dichte: 1.269 g/cm³ (22.5 °C) |
MDL:
MFCD00039232 CAS-Nummer: 500-73-2 EINECS: 207-911-4 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...