1-Iod-2,2,3,3-tetrafluorpropan

Lieferant: FLUOROCHEM
Achtung

003396-5G 003396-25G
FLUO003396-5GEA 63.9 EUR
FLUO003396-5G FLUO003396-25G
1-Iod-2,2,3,3-tetrafluorpropan
1-Iod-2,2,3,3-tetrafluorpropan
Formel: C₃H₃F₄I
Molecular Weight: 241,95 g/mol
MDL: MFCD00019033
CAS-Nummer: 679-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR