1-Iod-3,3,3-trifluorpropan

Lieferant: FLUOROCHEM

Synonyme: 1,1,1-Trifluor-3-iodpropan

001547-5G 001547-100G 001547-25G
FLUO001547-5GEA 55.4 EUR
FLUO001547-5G FLUO001547-100G FLUO001547-25G
1-Iod-3,3,3-trifluorpropan
1-Iod-3,3,3-trifluorpropan
Formel: C₃H₄F₃I
Molecular Weight: 223,96 g/mol
MDL: MFCD00038531
CAS-Nummer: 460-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR