1-Iod-4,4,4-trifluorbutan

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1,1,1-Trifluor-4-iodbutan

001539-5G 001539-1G 001539-25G
FLUO001539-5GEA 107 EUR
FLUO001539-5G FLUO001539-1G FLUO001539-25G
1-Iod-4,4,4-trifluorbutan
1-Iod-4,4,4-trifluorbutan
Formel: C₄H₆F₃I
Molecular Weight: 237,99 g/mol
MDL: MFCD00041543
CAS-Nummer: 461-17-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR