1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluor-8-iodnonan

Lieferant: FLUOROCHEM

Synonyme: 2-Iodo-1H,1H,1H,2H,3H,3H-perfluorononane

010550-1G 010550-5G
FLUO010550-1GEA 0 EUR
FLUO010550-1G FLUO010550-5G
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluor-8-iodnonan
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluor-8-iodnonan
Formel: C₉H₆F₁₃I
Molecular Weight: 488.03 g/mol
MDL: MFCD00156018
CAS-Nummer: 38550-34-4
EINECS: 253-994-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR