1-Amino-2,2,2-trifluorethan 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,2,2-Trifluorethylamin

B20789.03 B20789.06 B20789.14
B20789.03EA 33.4 EUR
B20789.03 B20789.06 B20789.14
1-Amino-2,2,2-trifluorethan 98%
1-Amino-2,2,2-trifluorethan

Purity: 98% CAS No.: 753-90-2 Molecular Formula: CF3CH2NH2 Formula Weight: 99.06 Storage Temperature: 2°C to 8°C Flash Point: -16°(3°F) Refractive Index: 1.2950 Odour: Amine-like Boiling Point: 36-38° Beilstein Reference: 1733204 UN No.: 2733 MDL No.: MFCD00008132

Formel: C₂H₄F₃N
Molecular Weight: 99,06 g/mol
Siedepunkt: 36…38 °C
Dichte: 1,245
Flammpunkt: –16 °C (3 °F)
MDL: MFCD00008132
CAS-Nummer: 753-90-2
EINECS: 212-041-3
UN: 2733
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR