1-Amino-2,2,2-trifluorethan
Lieferant: FLUOROCHEM
Synonyme:
2,2,2-Trifluorethylamin
003057-5G
003057-25G
003057-100G
FLUO003057-5GEA
55.4
EUR
FLUO003057-5G
FLUO003057-25G
FLUO003057-100G
1-Amino-2,2,2-trifluorethan
1-Amino-2,2,2-trifluorethan
Formel:
C₂H₄F₃N Molecular Weight: 99,06 g/mol Siedepunkt: 35,85 °C (1013 hPa) Dichte: 1,2452 g/cm³ (25 °C) |
MDL:
MFCD00008132 CAS-Nummer: 753-90-2 EINECS: 212-041-3 UN: 2733 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...