1-Amino-2,2,2-trifluorethan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,2,2-Trifluorethylamin
91692-25ML
91692-5ML
SIAL91692-25MLEA
838
EUR
SIAL91692-25ML
SIAL91692-5ML
1-Amino-2,2,2-trifluorethan, Sigma-Aldrich®
1-Amino-2,2,2-trifluorethan
Formel:
C₂H₄F₃N Molecular Weight: 99,06 g/mol Siedepunkt: 35,85 °C (1013 hPa) Dichte: 1,2452 g/cm³ (25 °C) |
MDL:
MFCD00008132 CAS-Nummer: 753-90-2 EINECS: 212-041-3 UN: 2733 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...