2,2,3,3,3-Pentafluor-1-propanol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
1H,1H-Pentafluor-1-propanol
001522-100G
001522-25G
FLUO001522-100GEA
195
EUR
FLUO001522-100G
FLUO001522-25G
2,2,3,3,3-Pentafluor-1-propanol
2,2,3,3,3-Pentafluor-1-propanol
Formel:
C₃H₃F₅O Molecular Weight: 150.05 g/mol Siedepunkt: 78…80 °C (1013 hPa) Dichte: 1.5 g/cm³ (20 °C) |
MDL:
MFCD00004673 CAS-Nummer: 422-05-9 EINECS: 207-012-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...