3,3,4,4,4-Pentafluor-1-butanol
Lieferant: FLUOROCHEM
010501-1G
010501-5G
FLUO010501-1GEA
57.9
EUR
FLUO010501-1G
FLUO010501-5G
3,3,4,4,4-Pentafluor-1-butanol
3,3,4,4,4-Pentafluor-1-butanol
Formel:
C₄H₅F₅O Molecular Weight: 164,08 g/mol |
MDL:
MFCD03424479 CAS-Nummer: 54949-74-5 EINECS: 259-405-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...