3,3,4,4,4-Pentafluor-2-butanol

Lieferant: FLUOROCHEM

006957-1G 006957-5G
FLUO006957-1GEA 69.8 EUR
FLUO006957-1G FLUO006957-5G
3,3,4,4,4-Pentafluor-2-butanol
3,3,4,4,4-Pentafluor-2-butanol
Formel: C₄H₅F₅O
Molecular Weight: 164,08 g/mol
Siedepunkt: 85 °C (1013 hPa)
Dichte: 1,406 g/cm³ (20 °C)
MDL: MFCD00042432
CAS-Nummer: 374-40-3
EINECS: 206-776-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR