Natriumtrifluormethansulfonat
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Trifluormethansulfonsäure Natriumsalz
20837.04
20837.09
20837.18
20837.04EA
31.3
EUR
20837.04
20837.09
20837.18
Natriumtrifluormethansulfonat
Natriumtrifluormethansulfonat
CAS No.: 2926-30-9
Molecular Formula: CF3NaO3S
Formula Weight: 172.06
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 253-255°
Beilstein Reference: 3728797
MDL No.: MFCD00061607
Formel:
CF₃NaO₃S Molecular Weight: 172,06 g/mol Schmelzpunkt: 253…255 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061607 CAS-Nummer: 2926-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...