Iodacetonitril

Lieferant: FLUOROCHEM

Synonyme: 2-Iodoacetonitrile

001086-25G 001086-100G
FLUO001086-25GEA 317 EUR
FLUO001086-25G FLUO001086-100G
Iodacetonitril
Iodacetonitril
Formel: C₂H₂IN
Molecular Weight: 166.95 g/mol
Siedepunkt: 182
Dichte: 2.3065 g/cm³ (20 °C)
MDL: MFCD00001886
CAS-Nummer: 624-75-9
EINECS: 210-860-0
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR