N,N-Dimethylformamiddiethylacetal, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,1-Diethoxytrimethylamin
M-Clarity™ Qualitätsstufe = MQ100
40252-100ML-F
SIAL40252-100ML-FEA
240
EUR
SIAL40252-100ML-F
N,N-Dimethylformamiddiethylacetal, Sigma-Aldrich®
N,N-Dimethylformamiddiethylacetal
Formel:
C₇H₁₇NO₂ Molecular Weight: 147,22 g/mol Siedepunkt: 130 °C (1013 hPa) Dichte: 0,86 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009227 CAS-Nummer: 1188-33-6 EINECS: 214-707-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...