N,N-Dimethylhexylamin 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N,N-Dimethylhexan-1-amine
A17364.06
A17364.14
A17364.22
A17364.06EA
59.3
EUR
A17364.06
A17364.14
A17364.22
N,N-Dimethylhexylamin 99%
N,N-Dimethylhexylamin
Purity: 99%
CAS No.: 4385-04-0
Molecular Formula: CH3(CH2)5N(CH3)2
Formula Weight: 129.25
Flash Point: 33°(92°F)
Refractive Index: 1.4136
Boiling Point: 146-150°
UN No.: 2733
MDL No.: MFCD00009522
Formel:
C₈H₁₉N Molecular Weight: 129,25 g/mol Siedepunkt: 146…150 °C Dichte: 0,744 Flammpunkt: 33 °C (92 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009522 CAS-Nummer: 4385-04-0 EINECS: 224-491-8 UN: 2733 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...