N,N-Dimethylhexylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N,N-Dimethylhexan-1-amine

308102-25G 308102-5G
SIAL308102-25GEA 211 EUR
SIAL308102-25G SIAL308102-5G
N,N-Dimethylhexylamin, Sigma-Aldrich®
N,N-Dimethylhexylamin
Formel: C₈H₁₉N
Molecular Weight: 129,25 g/mol
MDL: MFCD00009522
CAS-Nummer: 4385-04-0
EINECS: 224-491-8
UN: 2733
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR