N,N,N'-Trimethylethylendiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Dimethylamino)-2-(methylamino)ethanDimethyl(2-(methylamino)ethyl)amin

SIAL127124-5GEA 99.8 EUR
SIAL127124-5G SIAL127124-25G
N,N,N'-Trimethylethylendiamin, Sigma-Aldrich®
N,N,N'-Trimethylethylendiamin
Formel: C₅H₁₄N₂
Molecular Weight: 102,18 g/mol
Siedepunkt: 116 °C (1013 hPa)
Dichte: 0,783 g/cm³ (20 °C)
MDL: MFCD00014874
CAS-Nummer: 142-25-6
EINECS: 205-529-2
UN: 2733
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR